Learn More
Apexbio Technology LLC 5-Iodo-UTP(Synonyms: 5-Iodouridine triphosphate, 5-Iodo-UTP sodium salt, Iodo-UTP, 5-Iodo-uridine 5'-triphosphate), 10ul (100 mM).
SDP
Catalog No. 5000060795
Encompass_Preferred
5-Iodo-UTP is an iodinated derivative of uridine triphosphate (UTP) serving as a substrate in RNA synthesis and modification reactions In vitro RNA polymerases incorporate 5-Iodo-UTP into RNA transcripts enabling the production of labeled RNA probes for hybridization assays nucleic acid localization studies and analyses of RNA-protein interactions It also facilitates the investigation of RNA polymerase substrate specificity and the functional mapping of RNA molecules
Catalog No. 50-000-60795 Supplier Apexbio Technology LLC Supplier No. B79650.01
5-Iodo-UTP is an iodinated derivative of uridine triphosphate (UTP) serving as a substrate in RNA synthesis and modification reactions In vitro RNA polymerases incorporate 5-Iodo-UTP into RNA transcripts enabling the production of labeled RNA probes for hybridization assays nucleic acid localization studies and analyses of RNA-protein interactions It also facilitates the investigation of RNA polymerase substrate specificity and the functional mapping of RNA molecules