Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Iodoindole, 98%

Catalog No. AAB2063814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2063814 25 g
AAB2063806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2063814 Supplier Thermo Scientific Chemicals Supplier No. B2063814
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 16066-91-4 | C8H6IN | 243.047 g/mol

5-Iodoindole is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. It is used as the starting material in the synthesis of 5-(α-fluorovinyl)-N-tosylindole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Iodoindole is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. It is used as the starting material in the synthesis of 5-(α-fluorovinyl)-N-tosylindole.

Notes
Store at room temperature. Incompatible with strong oxidizing agents. Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16066-91-4
Molecular Formula C8H6IN
Molecular Weight (g/mol) 243.047
MDL Number MFCD00220065
InChI Key TVQLYTUWUQMGMP-UHFFFAOYSA-N
Synonym 5-iodoindole, 1h-indole, 5-iodo, 5-iodo indole, 5-iodanyl-1h-indole, zlchem 1099, acmc-1brxb, maybridge1_006499
PubChem CID 2782313
IUPAC Name 5-iodo-1H-indole
SMILES C1=CC2=C(C=CN2)C=C1I

Specifications

Melting Point 97°C to 100°C
Quantity 25 g
Sensitivity Light sensitive
Formula Weight 243.05
Percent Purity 98%
Chemical Name or Material 5-Iodoindole
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.