Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Iodoisatin, MP Biomedicals

Catalog No. ICN212521 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN212521 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN212521 Supplier MP Biomedicals, Inc Supplier No. 0521252105

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20780-76-1
Molecular Formula C8H4INO2
Molecular Weight (g/mol) 273.029
InChI Key OEUGDMOJQQLVAZ-UHFFFAOYSA-N
Synonym 5-iodoisatin, 5-iodoindoline-2,3-dione, 5-iodo isatin, 5-iodoindole-2,3-dione, 5-iodo-2,3-dioxoindoline, 1h-indole-2,3-dione, 5-iodo, 5-iodo-2,3-dihydro-1h-indole-2,3-dione, 5-iodo-1h-benzo d azolidine-2,3-dione, 5-iodo-isatine, 5-iodo-isatin
PubChem CID 88695
IUPAC Name 5-iodo-1H-indole-2,3-dione
SMILES C1=CC2=C(C=C1I)C(=O)C(=O)N2

Specifications

Quantity 5 g
Formula Weight 273.03
Chemical Name or Material 5-Iodoisatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.