Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-iodopicolinic acid | 32046-43-8 | MFCD09910324 | 249.01 g/mol | C6H4INO2 | 1 G
SDP

Catalog No. 5000208795
Encompass_Preferred

5-Iodopicolinic acid (5-iodopyridine-2-carboxylic acid) is an iodinated pyridine carboxylic acid used as a chemical intermediate in organic synthesis and drug research. It serves as a building block for halogenated ligands and coordination complexes.

  • iodinated pyridine carboxylic acid
  • molecular formula C6H4INO2
  • molecular weight 249.01 g/mol
  • cas number 32046-43-8
  • intended use as a drug intermediate
  • store under conditions specified in the certificate of analysis
  • for research use only; not for human or clinical use

Catalog No. 50-002-08795 Supplier Medchemexpress LLC Supplier No. HYW0416571G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Iodopicolinic acid (5-iodopyridine-2-carboxylic acid) is an iodinated pyridine carboxylic acid used as a chemical intermediate in organic synthesis and drug research. It serves as a building block for halogenated ligands and coordination complexes.

  • iodinated pyridine carboxylic acid
  • molecular formula C6H4INO2
  • molecular weight 249.01 g/mol
  • cas number 32046-43-8
  • intended use as a drug intermediate
  • store under conditions specified in the certificate of analysis
  • for research use only; not for human or clinical use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.