Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-iodopyridine-2-carboxylic acid | 32046-43-8 | MFCD09910324 | 95.0% | 249.01 g·mol^-1 | C6H4INO2 | 250 MG
SDP

Catalog No. 5000209443
Encompass_Preferred

5-Iodopicolinic acid (5-iodopyridine-2-carboxylic acid) is an iodinated pyridine carboxylic acid used primarily as a synthetic intermediate in organic and medicinal chemistry. It is supplied for research and analytical applications and is commonly used to prepare halogenated derivatives and other active compounds.

  • Used as a synthetic intermediate in organic and medicinal chemistry.
  • Useful for synthesis of halogenated derivatives and active compounds.
  • Available in multiple package sizes, including 250 mg.
  • Typical appearance: yellow to off-white solid.
  • Molecular formula C6H4INO2 and molecular weight 249.01 g·mol^-1.
  • CAS number 32046-43-8 for unambiguous identification.

Catalog No. 50-002-09443 Supplier Medchemexpress LLC Supplier No. 250MGHYW041657
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Iodopicolinic acid (5-iodopyridine-2-carboxylic acid) is an iodinated pyridine carboxylic acid used primarily as a synthetic intermediate in organic and medicinal chemistry. It is supplied for research and analytical applications and is commonly used to prepare halogenated derivatives and other active compounds.

  • Used as a synthetic intermediate in organic and medicinal chemistry.
  • Useful for synthesis of halogenated derivatives and active compounds.
  • Available in multiple package sizes, including 250 mg.
  • Typical appearance: yellow to off-white solid.
  • Molecular formula C6H4INO2 and molecular weight 249.01 g·mol^-1.
  • CAS number 32046-43-8 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.