Learn More
Enzo Life Sciences 5-Iodotubercidin (5mg). CAS: 24386-93-4
SDP
Supplier:  Enzo Life Sciences BMLEI2930005
Encompass_Preferred
Inhibits ERK2 (Ki = 525nM). Also inhibits adenosine kinase (Ki = 30nM), CK1 and CK2 and insulin receptor kinase (IC50 ranging from 0.4-28µM). Alternative name: 4-Amino-5-iodo-7-(beta-D-ribofuranosyl)-pyrrolo 2 3-d pyrimidine. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (8mg/ml warm). Long Term Storage: -20°C.
Catalog No. 50-201-0675