Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Iodouracil

Catalog No. 85785810G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-785-810G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 85-785-810G Supplier Sigma Aldrich Supplier No. 85785810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Building block has been shown to be used as a key intermediate in the synthesis of memantine hydrochloride, which was reported to be a FDA approved drug for Alzheimer's treatment.An Improved Synthesis of Memantine Hydrochloride: Anti-Alzheimer's Drug

Specifications

CAS 696-07-1
Melting Point 274°C to 276°C (dec.) (lit.)
Molecular Formula C4H3IN2O2
Linear Formula C4H3IN2O2
MDL Number MFCD00006020
Quantity 250 mg
Synonym 2,4-Dihydroxy-5-iodopyrimidine
Molecular Weight (g/mol) 237.98
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.