Learn More
Cayman Chemical 5-IPF2a-VI 10ug
SDP
Catalog No. 502972043
Encompass_Preferred
A psychoactive analog of p-iodoamphetamine that is indistinguishable in its physiological effects from MDMA in rats significantly reduces both serotonin uptake sites and hippocampal serotonin levels in rats intended for forensic applications
Catalog No. 50-297-2043 Supplier Cayman Chemical Supplier No. 1630010 UG
A psychoactive analog of p-iodoamphetamine that is indistinguishable in its physiological effects from MDMA in rats significantly reduces both serotonin uptake sites and hippocampal serotonin levels in rats intended for forensic applications