Learn More
Cayman Chemical 5-IPF2a-VI 25ug
SDP
Catalog No. 502972624
Encompass_Preferred
A water-soluble derivative of geldanamycin which potently inhibits Hsp90 (IC50 24 nM) and has excellent bioavailability and tissue distribution in animals has diverse anti-tumor actions suppresses inflammation by interfering with signaling through the NF-KB pathway ameliorates high fat diet-induced renal failure
Catalog No. 50-297-2624 Supplier Cayman Chemical Supplier No. 1630025 UG
A water-soluble derivative of geldanamycin which potently inhibits Hsp90 (IC50 24 nM) and has excellent bioavailability and tissue distribution in animals has diverse anti-tumor actions suppresses inflammation by interfering with signaling through the NF-KB pathway ameliorates high fat diet-induced renal failure