Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Isopropylamino)pentanol 98.0+%, TCI America™
SDP

Catalog No. I07571G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I07571G Supplier TCI America Supplier No. I07571G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40447-21-0
Molecular Formula C8H19NO
Molecular Weight (g/mol) 145.246
InChI Key UEXRIBOEINFZJS-UHFFFAOYSA-N
PubChem CID 414092
IUPAC Name 5-(propan-2-ylamino)pentan-1-ol
SMILES CC(C)NCCCCCO

Specifications

Melting Point 44°C
Color White
Boiling Point 125°C
Quantity 1 g
Formula Weight 145.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(Isopropylamino)pentanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.