Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Isoxazol-5-ylthiophene-2-sulfonyl chloride, Tech., Thermo Scientific™

Catalog No. CC16003DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC16003DE 5 g
CC16003CB 250 mg
CC16003DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC16003DE Supplier Thermo Scientific Chemicals Supplier No. CC16003DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 551930-53-1
Molecular Formula C7H4ClNO3S2
Molecular Weight (g/mol) 249.68
InChI Key SSGKBJJYLTYNQD-UHFFFAOYSA-N
Synonym 5-5-isoxazyl thiophene-2-sulfonyl chloride, 5-isoxazol-5-ylthiophene-2-sulfonyl chloride, 5-1,2-oxazol-5-yl thiophene-2-sulfonyl chloride, 5-5-isoxazolyl-2-thiophenesulfonyl chloride, 5-isoxazol-5-yl thiophene-2-sulfonyl chloride, chloro 5-isoxazol-5-yl 2-thienyl sulfone, ssgkbjjyltynqd-uhfffaoysa-n, 5-5-isoxazyl thiophene-2-sulphonyl chloride, 5-5-isoxazolyl-2-thiophenesulfonoyl chloride, 5-5-isoxazolyl thiophene-2-sulfonyl chloride
PubChem CID 2795330

Specifications

Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 249.7
Percent Purity Tech.
Grade Technical
Chemical Name or Material 5-Isoxazol-5-ylthiophene-2-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.