Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-isoxazoleacetic acid, a-amino-3-chloro-4,5-dihydro-, monohydrochloride, [S-(R*,R*)]- | 161922-40-3 | MFCD32874156 | 99.1% | 215.03 g·mol⁻¹ | C5H8Cl2N2O3 | 25 MG
SDP

Catalog No. 5000303494
Encompass_Preferred

Acivicin hydrochloride is the hydrochloride salt of acivicin, a natural product γ-glutamyl transpeptidase (GGT) inhibitor used as a biochemical research reagent. It is supplied as a solid and in solution formats for laboratory use, with defined storage conditions to preserve stability and activity.

  • High confirmed purity (HPLC): 99.1%.
  • Molecular weight 215.03 g·mol⁻¹ and formula C5H8Cl2N2O3.
  • Available in mg-scale packs and solution aliquots for assay flexibility.
  • Suitable for biochemical studies targeting GGT and related pathways.
  • Storage recommendations: 4°C under nitrogen; in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-003-03494 Supplier Medchemexpress LLC Supplier No. 25MGHYW016586A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acivicin hydrochloride is the hydrochloride salt of acivicin, a natural product γ-glutamyl transpeptidase (GGT) inhibitor used as a biochemical research reagent. It is supplied as a solid and in solution formats for laboratory use, with defined storage conditions to preserve stability and activity.

  • High confirmed purity (HPLC): 99.1%.
  • Molecular weight 215.03 g·mol⁻¹ and formula C5H8Cl2N2O3.
  • Available in mg-scale packs and solution aliquots for assay flexibility.
  • Suitable for biochemical studies targeting GGT and related pathways.
  • Storage recommendations: 4°C under nitrogen; in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.