Learn More
Analytical Sales & Services 5µL PEEK Sample Loop
Catalog No. 5000807210
Encompass
5µL PEEK Sample Loop for High Pressure Automatic Switching Valves and 9125, 9725 and 9010 PEEK Manual Injector Valves\n\nQty/Pkg: EACH
Catalog No. 50-008-07210 Supplier Analytical Sales & Services Supplier No. 490550204
5µL PEEK Sample Loop for High Pressure Automatic Switching Valves and 9125, 9725 and 9010 PEEK Manual Injector Valves\n\nQty/Pkg: EACH