Learn More
Perkin Elmer US LLC 5 µL Syringe, Metal Plunger PTFE-tipped Seal 0.63 mm OD Needle BLUE Barrel (10/pack)
Catalog No. 502111524
Encompass_Preferred
Syringes from PerkinElmer are individually inspected for accuracy and performance. Recommended autosampler syringes are available in 0.5, 5 and 50 µL capacities. For routine analyses, the metal plunger in barrel with PTFE-tipped seal is the standard syringe as shipped with each Clarus GC instrument. Alternative syringes to use are the metal plunger in barrel or the 0.53 mm on-column injection.
Catalog No. 50-211-1524 Supplier Perkin Elmer US LLC Supplier No. N6103241
Syringes from PerkinElmer are individually inspected for accuracy and performance. Recommended autosampler syringes are available in 0.5, 5 and 50 µL capacities. For routine analyses, the metal plunger in barrel with PTFE-tipped seal is the standard syringe as shipped with each Clarus GC instrument. Alternative syringes to use are the metal plunger in barrel or the 0.53 mm on-column injection.