Learn More
Alkali Scientific CellPro™ 5-Layer Cell Culture Multi-Layer Flasks, 875cm², Vent Cap, TC treated, Sterile, 1/pk, 8/cs
SDP
Catalog No. NC1973866
Encompass_Preferred
Multi-Layer, 5-Layer Cell Culture Flask provide 870 cm2 cell growth surface area. The tissue culture treated Multi-layer Flasks enable you to grow more cells faster and easier, thereby making your cell culture workflow more productive, easier, and in a more convenient manner. Vacuum plasma TC treatment for the best cell attachment. Sterilized by E-Beam. Non-Pyrogenic and DNase/RNase free.
Catalog No. NC1973866 Supplier Alkali Scientific Supplier No. TV55L
Multi-Layer, 5-Layer Cell Culture Flask provide 870 cm2 cell growth surface area. The tissue culture treated Multi-layer Flasks enable you to grow more cells faster and easier, thereby making your cell culture workflow more productive, easier, and in a more convenient manner. Vacuum plasma TC treatment for the best cell attachment. Sterilized by E-Beam. Non-Pyrogenic and DNase/RNase free.