Learn More
Thomas Scientific 5-Layer Cell Culture Flask, Vent Cap, Straight Neck, TC, Sterile
SDP
Catalog No. 501461375
Encompass_Preferred
Nest Multi-Layer Flasks provide 870 cm2 cell growth surface area. The tissue culture treated NEST Multi-layer Flasks enable you to grow more cells faster and easier, thereby making your cell culture workflow more productive, easier, and in a more convenient manner. Vacuum plasma TC treatment for the best cell attachment Sterilized by E-Beam Non-Pyrogenic, DNase/RNase FREE Both plug seal cap and vent cap are available
This item is not returnable. View return policy
Catalog No. 50-146-1375 Supplier Thomas Scientific Supplier No. 1194Z19
Nest Multi-Layer Flasks provide 870 cm2 cell growth surface area. The tissue culture treated NEST Multi-layer Flasks enable you to grow more cells faster and easier, thereby making your cell culture workflow more productive, easier, and in a more convenient manner. Vacuum plasma TC treatment for the best cell attachment Sterilized by E-Beam Non-Pyrogenic, DNase/RNase FREE Both plug seal cap and vent cap are available