Learn More
Bellco Glass, Inc. 5-Layer Cell Culture Flask, Vent Cap, Straight Neck, TC, sterile 1/pk, 8/cs
SDP
Supplier:  Bellco Glass, Inc. 451400076/SQ1026867
Encompass_Preferred
5-Layer Cell Culture Flask, Plug Seal Cap, Straight Neck, TC, sterile 1/pk, 8/cs. Multi-Layer Flasks provide 870 cm2 cell growth surface area. The tissue culture treated Multi-layer Flasks enable you to grow more cells faster and easier, thereby making your cell culture workflow more productive, easier, and in a more convenient manner. • Made of high clarity, 100% virgin polystyrene.• Sterilized by E-beam, SAL=10-6.• Non-Pyrogenic, DNase/Rnase free.• Growth area: 870 cm².• Individually Packaged in sterile bag.
Catalog No. NC3865183