Learn More
Chemglass Life Sciences Chemglass 5 L Expansion tank for Huber CC505 and CC508
SDP
Catalog No. NC3457160
Encompass
The Chemglass 5 L Expansion Tank (CG-1969-T05) provides additional fluid capacity for temperature control units to accommodate thermal expansion of heat transfer fluids. Designed for use with Huber CC505 and CC508 circulators, it is intended for applications where the unit's onboard reservoir is insufficient.
  • Provides 5 L additional fluid capacity to accommodate thermal expansion of heat transfer fluids.
  • Reduces risk of overpressure and improves system stability during high-temperature operation.
  • Intended for use when the temperature control unit's onboard capacity is insufficient.
  • Consult technical services for compatibility confirmation and quotation.
Catalog No. NC3457160 Supplier Chemglass Life Sciences Supplier No. CG1969T05
The Chemglass 5 L Expansion Tank (CG-1969-T05) provides additional fluid capacity for temperature control units to accommodate thermal expansion of heat transfer fluids. Designed for use with Huber CC505 and CC508 circulators, it is intended for applications where the unit's onboard reservoir is insufficient.
  • Provides 5 L additional fluid capacity to accommodate thermal expansion of heat transfer fluids.
  • Reduces risk of overpressure and improves system stability during high-temperature operation.
  • Intended for use when the temperature control unit's onboard capacity is insufficient.
  • Consult technical services for compatibility confirmation and quotation.