Learn More
Chemglass Life Sciences 5000mL Glas-Col Heating Mantle, Series TM, Spherical, 600 Watts
SDP
Catalog No. CG1200011
Encompass_Preferred
5000mL Glas-Col Heating Mantle, Series TM, Spherical, 600 Watts. The Glas-Col Series TM spherical flask mantle covers only the flasks bottom half, letting you see its full contents. Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full.The Series TM mantle is available with a bottom hole for flask drain on a special order basis. Terminal box models are available with switches and pilot lights. Mantles are 115 Volt.
Catalog No. CG1200011 Supplier Chemglass Life Sciences Supplier No. CG1200011
5000mL Glas-Col Heating Mantle, Series TM, Spherical, 600 Watts. The Glas-Col Series TM spherical flask mantle covers only the flasks bottom half, letting you see its full contents. Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full.The Series TM mantle is available with a bottom hole for flask drain on a special order basis. Terminal box models are available with switches and pilot lights. Mantles are 115 Volt.