Learn More
AdipoGen 5-Maleimido-eosin
SDP
Supplier:  AdipoGen CDXM0013M010
Encompass_Preferred
Chemical. CAS 150322-02-4. Formula C24H9Br4NO7. MW 742.95. Synthetic. Triplet probe used for the measurment of rotational diffusion of protein in solution and in membranes, includin myosin and pyruvate dehydrogenase.
Catalog No. 50-256-0576