Learn More
Medchemexpress LLC 5-maleimido-pentanoic N-hydroxysuccinimide ester | 103750-03-4 | MFCD01318603 | 99.3% | 294.26 g/mol | C13H14N2O6 | 50 MG
SDP
Catalog No. 5000202443
Encompass_Preferred
Mal-C2-NHS ester (5-maleimido-pentanoic N-hydroxysuccinimide ester) is a noncleavable antibody-drug conjugate (ADC) linker that features a maleimide and an N-hydroxysuccinimide (NHS) ester for bioconjugation to thiol and amine groups during ADC synthesis.
  • Noncleavable ADC linker suitable for antibody-drug conjugation.
  • Maleimide group enables selective thiol coupling.
  • N-hydroxysuccinimide (NHS) ester enables amine coupling.
  • High purity and defined molecular weight for reproducible conjugation.
Catalog No. 50-002-02443 Supplier Medchemexpress LLC Supplier No. HY12650250MG
Mal-C2-NHS ester (5-maleimido-pentanoic N-hydroxysuccinimide ester) is a noncleavable antibody-drug conjugate (ADC) linker that features a maleimide and an N-hydroxysuccinimide (NHS) ester for bioconjugation to thiol and amine groups during ADC synthesis.
  • Noncleavable ADC linker suitable for antibody-drug conjugation.
  • Maleimide group enables selective thiol coupling.
  • N-hydroxysuccinimide (NHS) ester enables amine coupling.
  • High purity and defined molecular weight for reproducible conjugation.