Learn More
Abcam 5-Maleimidoeosin, Fluorescent triplet probe, 10MG
Catalog No. 502705267
Encompass_Preferred
MW 742.95 g/mol, Purity >93%. Fluorescent triplet probe used for the measurement of rotational diffusion of protein in solution and in membranes, including myosin and pyruvate dehydrogenase. Widely used in a flow cytometry test to detect hereditary spherocytosis (HS).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5267 Supplier Abcam Supplier No. AB14535510MG
MW 742.95 g/mol, Purity >93%. Fluorescent triplet probe used for the measurement of rotational diffusion of protein in solution and in membranes, including myosin and pyruvate dehydrogenase. Widely used in a flow cytometry test to detect hereditary spherocytosis (HS).
The product is subject to the following: Abcam Restricted Use Statement