Learn More
Medchemexpress LLC 5-maleimidovaleric acid | 57078-99-6 | 98.0% | 197.19 g/mol | C9H11NO4 | 250 MG
SDP
Catalog No. 5000202680
Encompass_Preferred
5-Maleimidovaleric acid is a cleavable antibody-drug conjugate (ADC) linker used for conjugation via maleimide-thiol chemistry. Supplied as an off-white to yellow solid reagent for research use in ADC synthesis and related conjugation reactions. Typical purity is 98.0%, molecular formula C9H11NO4, and molecular weight 197.19 g/mol. Store powdered material at -20°C for long-term stability.
  • Cleavable ADC linker for maleimide-thiol conjugation.
  • Off-white to yellow solid with 98.0% purity.
  • Molecular formula C9H11NO4; molecular weight 197.19 g/mol.
  • Available in multiple pack sizes including 250 mg.
  • Intended for research use only; not for human use.
Catalog No. 50-002-02680 Supplier Medchemexpress LLC Supplier No. HY140987250MG
5-Maleimidovaleric acid is a cleavable antibody-drug conjugate (ADC) linker used for conjugation via maleimide-thiol chemistry. Supplied as an off-white to yellow solid reagent for research use in ADC synthesis and related conjugation reactions. Typical purity is 98.0%, molecular formula C9H11NO4, and molecular weight 197.19 g/mol. Store powdered material at -20°C for long-term stability.
  • Cleavable ADC linker for maleimide-thiol conjugation.
  • Off-white to yellow solid with 98.0% purity.
  • Molecular formula C9H11NO4; molecular weight 197.19 g/mol.
  • Available in multiple pack sizes including 250 mg.
  • Intended for research use only; not for human use.