Learn More
Cayman Chemical 5-MAPB (hydrochloride), Purity: ≥98%, CAS: 1823925-53-6, 10 mg
SDP
Catalog No. 502953411
Encompass_Preferred
An analytical reference standard categorized as a benzofuran; has been found in seized samples suspected of containing NPS; intended for research and forensic applications
Catalog No. 50-295-3411 Supplier Cayman Chemical Supplier No. 1410910 MG
An analytical reference standard categorized as a benzofuran; has been found in seized samples suspected of containing NPS; intended for research and forensic applications