Learn More
Zymo Research Corporation 5-mC DNA ELISA Kit, 1 x 96 Preps
SDP
Supplier:  Zymo Research Corporation D5325
Encompass_Preferred
The kit features a unique Anti-5-Methylcytosine monoclonal antibody that is both sensitive and specific for 5-mC. The assay is compatible with a wide range of input DNA from vertebrate, plant, and microbial sources as well as PCR amplicons and fragmented DNA. Percent 5-mC in a DNA sample can be accurately quantified from a standard curve generated with specially designed controls included with the kit. Also, the fast, streamlined workflow is ideal for high-throughput analyses.
Catalog No. 50-444-393