Learn More
Medchemexpress LLC 5-Me-dC(Ac) amidite | 1873306-74-1 | MFCD11114390 | 97.0% | 785.86 | C42H52N5O8P | 1 ML
SDP
Catalog No. 5000284891
Encompass_Preferred
5-Me-dC(Ac) amidite is a nucleoside phosphoramidite for solid-phase synthesis of modified DNA and RNA oligonucleotides that installs an N4-acetyl-5-methyl-deoxycytidine residue. Available as a ready-to-use solution or as solid material for reconstitution. CAS 1873306-74-1; formula C42H52N5O8P; MW 785.86.
  • Designed for incorporation of N4-acetyl-5-methyl-deoxycytidine during oligonucleotide synthesis.
  • Available as a ready-to-use 10 mM solution in DMSO or as solid for custom reconstitution.
  • High solubility in DMSO (100 mg/mL) with ultrasonic assistance.
  • Storage recommendations: solids at 4°C protected from light; solutions -80°C up to 6 months, -20°C up to 1 month.
  • Suitable for studies of DNA methylation and epigenetic modification effects.
Catalog No. 50-002-84891 Supplier Medchemexpress LLC Supplier No. HYW57088410MM/1ML
5-Me-dC(Ac) amidite is a nucleoside phosphoramidite for solid-phase synthesis of modified DNA and RNA oligonucleotides that installs an N4-acetyl-5-methyl-deoxycytidine residue. Available as a ready-to-use solution or as solid material for reconstitution. CAS 1873306-74-1; formula C42H52N5O8P; MW 785.86.
  • Designed for incorporation of N4-acetyl-5-methyl-deoxycytidine during oligonucleotide synthesis.
  • Available as a ready-to-use 10 mM solution in DMSO or as solid for custom reconstitution.
  • High solubility in DMSO (100 mg/mL) with ultrasonic assistance.
  • Storage recommendations: solids at 4°C protected from light; solutions -80°C up to 6 months, -20°C up to 1 month.
  • Suitable for studies of DNA methylation and epigenetic modification effects.