Learn More
Medchemexpress LLC 5-Me-dC(Ac) amidite | 1873306-74-1 | MFCD11114390 | 97.0% | 785.86 | C42H52N5O8P | 100 MG
SDP
Catalog No. 5000283990
Encompass_Preferred
5-Me-dC(Ac) amidite is a protected 5-methyl-2′-deoxycytidine phosphoramidite reagent designed for incorporation of 5-methylcytosine into synthetic DNA or RNA during automated solid-phase oligonucleotide synthesis. Supplied for research use, the product is provided as solids or as ready-to-use solutions to simplify synthesis workflows.
  • Protected 5-methylcytosine phosphoramidite for site-specific incorporation.
  • High purity (97.0%) suitable for oligonucleotide synthesis.
  • Available as solid and 10 mM solution in DMSO for immediate use.
  • Pack sizes include 50 mg, 100 mg, 250 mg, and 1 g; larger quantities available by quote.
  • Documentation provided: datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-83990 Supplier Medchemexpress LLC Supplier No. HYW570884100MG
5-Me-dC(Ac) amidite is a protected 5-methyl-2′-deoxycytidine phosphoramidite reagent designed for incorporation of 5-methylcytosine into synthetic DNA or RNA during automated solid-phase oligonucleotide synthesis. Supplied for research use, the product is provided as solids or as ready-to-use solutions to simplify synthesis workflows.
  • Protected 5-methylcytosine phosphoramidite for site-specific incorporation.
  • High purity (97.0%) suitable for oligonucleotide synthesis.
  • Available as solid and 10 mM solution in DMSO for immediate use.
  • Pack sizes include 50 mg, 100 mg, 250 mg, and 1 g; larger quantities available by quote.
  • Documentation provided: datasheet, certificate of analysis, and safety data sheet.