Learn More
Medchemexpress LLC 5-Me-dC(Ac) amidite (5-methyl-2'-deoxycytidine phosphoramidite) | 1873306-74-1 | MFCD11114390 | 97.0% | 785.86 | C42H52N5O8P | 1 G
SDP
Catalog No. 5000284980
Encompass_Preferred
5-Me-dC(Ac) amidite is a nucleoside phosphoramidite reagent used to introduce N4-acetyl-5-methyl-2'-deoxycytidine into synthetic DNA or RNA during automated solid-phase oligonucleotide synthesis. It is supplied as a solid and includes guidance for solubility and storage for research applications.
  • Enables incorporation of N4-acetyl-5-methyl-2'-deoxycytidine into oligonucleotides.
  • High purity suitable for synthesis and analytical applications.
  • Soluble in DMSO; compatible with common deprotection protocols.
  • Available in multiple sizes including 1 g for preparative use.
  • Supplied with datasheet and certificate of analysis for quality control.
  • Store protected from light; follow recommended solvent storage conditions.
Catalog No. 50-002-84980 Supplier Medchemexpress LLC Supplier No. HYW5708841G
5-Me-dC(Ac) amidite is a nucleoside phosphoramidite reagent used to introduce N4-acetyl-5-methyl-2'-deoxycytidine into synthetic DNA or RNA during automated solid-phase oligonucleotide synthesis. It is supplied as a solid and includes guidance for solubility and storage for research applications.
  • Enables incorporation of N4-acetyl-5-methyl-2'-deoxycytidine into oligonucleotides.
  • High purity suitable for synthesis and analytical applications.
  • Soluble in DMSO; compatible with common deprotection protocols.
  • Available in multiple sizes including 1 g for preparative use.
  • Supplied with datasheet and certificate of analysis for quality control.
  • Store protected from light; follow recommended solvent storage conditions.