Learn More
Medchemexpress LLC 5-Me-DMT-2'-O-Me-C(Bz)-CE-phosphoramidite | 166593-57-3 | MFCD15145377 | 98.0% | 877.96 g/mol | C48H56N5O9P | 250 MG
SDP
Catalog No. 5000282553
Encompass_Preferred
5-Me-DMT-2'-O-Me-C(Bz)-CE-phosphoramidite is a phosphoramidite monomer used in solid-phase oligonucleotide synthesis to incorporate 5-methyl-2'-O-methylcytidine residues with N4-benzoyl protection. Supplied as a white to off-white solid at high purity, it is intended for research use in nucleic acid chemistry, epigenetics studies, and aptamer development.
  • High purity suitable for research use.
  • Compatible with standard solid-phase DNA/RNA synthesis workflows.
  • Introduces 5-methyl-2'-O-methylcytidine with benzoyl protection.
  • White to off-white solid appearance for easy handling.
  • Stable powder with defined cold-storage recommendations.
  • Available in small and bulk sizes for scale flexibility.
Catalog No. 50-002-82553 Supplier Medchemexpress LLC Supplier No. HYW336559250MG
5-Me-DMT-2'-O-Me-C(Bz)-CE-phosphoramidite is a phosphoramidite monomer used in solid-phase oligonucleotide synthesis to incorporate 5-methyl-2'-O-methylcytidine residues with N4-benzoyl protection. Supplied as a white to off-white solid at high purity, it is intended for research use in nucleic acid chemistry, epigenetics studies, and aptamer development.
  • High purity suitable for research use.
  • Compatible with standard solid-phase DNA/RNA synthesis workflows.
  • Introduces 5-methyl-2'-O-methylcytidine with benzoyl protection.
  • White to off-white solid appearance for easy handling.
  • Stable powder with defined cold-storage recommendations.
  • Available in small and bulk sizes for scale flexibility.