Learn More
Medchemexpress LLC 5-Methoxy-2-benzimidazolethiol | 37052-78-1 | 180.23 | 100 G
SDP
Catalog No. 5000447349
Encompass_Preferred
5-Methoxy-2-benzimidazolethiol is a benzimidazole. The iodimetric determination of 5-Methoxy-2-benzimidazolethiol in alkaline media is studied.
  • For research use only.
  • Available in various sizes from 10 mM * 1 mL in DMSO solution to 100 g solid.
  • Multiple dissolution protocols are provided for in vivo experiments, using co-solvents like DMSO, PEG300/PEG400, Tween 80, and SBE-β-CD in Saline.
Catalog No. 50-004-47349 Supplier Medchemexpress LLC Supplier No. HYW008378100G
5-Methoxy-2-benzimidazolethiol is a benzimidazole. The iodimetric determination of 5-Methoxy-2-benzimidazolethiol in alkaline media is studied.
  • For research use only.
  • Available in various sizes from 10 mM * 1 mL in DMSO solution to 100 g solid.
  • Multiple dissolution protocols are provided for in vivo experiments, using co-solvents like DMSO, PEG300/PEG400, Tween 80, and SBE-β-CD in Saline.