Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methoxy-2-methylbenzoselenazole, TCI America™
SDP

Catalog No. M19005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M19005G Supplier TCI America Supplier No. M19005G

missing translation for 'validMainframeMsg'

Specifications

CAS 2946-17-0
Color Yellow
Boiling Point 142°C
Molecular Formula C9H9NOSe
Quantity 5 g
MDL Number MFCD00005558
UN Number 3283
Synonym 5-methoxy-2-methylbenzoselenazole, benzoselenazole, 5-methoxy-2-methyl, 5-methoxy-2-methylbenzselenazole, 2-methyl-5-methoxybenzoselenazole, 5-methoxy-2-methylbenzo d 1,3-selenazole, #, 5-methoxy-2-methylbenzo d 1,3 selenazole, 5-methoxy-2-methylbenzo d 1,3-selenazoline
InChI Key LVIYTMFDCXRSDL-UHFFFAOYSA-N
SMILES CC1=NC2=C([Se]1)C=CC(=C2)OC
IUPAC Name 5-methoxy-2-methyl-1,3-benzoselenazole
Molecular Weight (g/mol) 226.148
PubChem CID 76274
Formula Weight 226.14
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Methoxy-2-methylbenzoselenazole
Show More Show Less
EINECSNumber (5)-0498
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.