Learn More
eMolecules​ 1094072-17-9 | 5-Methoxy-2-(tributylstannyl)pyridine | Synthonix - Stock | MFCD10699152 | 398.178 | C18H33NOSn | 95.000 | CCCC[Sn](CCCC)(CCCC)c1ccc(OC)cn1 | 25g | 525920823
Catalog No. 502696681 Shop All eMolecules Inc. Products
Encompass_Preferred
5-Methoxy-2-(tributylstannyl)pyridine | Synthonix - Stock | 1094072-17-9 | MFCD10699152 | 398.178 | C18H33NOSn | 95.000 | CCCC[Sn](CCCC)(CCCC)c1ccc(OC)cn1 | 25g | 525920823
Catalog No. 50-269-6681 Supplier eMolecules​ Supplier No. 525920823
5-Methoxy-2-(tributylstannyl)pyridine | Synthonix - Stock | 1094072-17-9 | MFCD10699152 | 398.178 | C18H33NOSn | 95.000 | CCCC[Sn](CCCC)(CCCC)c1ccc(OC)cn1 | 25g | 525920823