Learn More
Medchemexpress LLC 5-methoxy-5-oxopentanoic acid | 1501-27-5 | MFCD00004409 | 97.0% | 146.14 | C6H10O4 | 1 ML
SDP
Catalog No. 5000300190
Encompass_Preferred
5-methoxy-5-oxopentanoic acid is an endogenous metabolite (CAS 1501-27-5) provided for research and analytical use. The compound has molecular formula C6H10O4 and molecular weight 146.14 g/mol. It is offered as ready-to-use 10 mM solutions in DMSO (1 mL) and as bulk solid quantities. Appearance is colorless to light yellow with a reported density of 1.164 g/cm3.
  • Endogenous metabolite useful as a reference standard for metabolic studies.
  • Available as ready-to-use 10 mM solution in DMSO for rapid use.
  • Also available in multiple powder sizes for bulk applications.
  • Documentation available including datasheet, SDS, COA, and spectral data.
  • Colorless to light yellow liquid with reported density 1.164 g/cm3.
Catalog No. 50-003-00190 Supplier Medchemexpress LLC Supplier No. HYW01752310MM/1ML
5-methoxy-5-oxopentanoic acid is an endogenous metabolite (CAS 1501-27-5) provided for research and analytical use. The compound has molecular formula C6H10O4 and molecular weight 146.14 g/mol. It is offered as ready-to-use 10 mM solutions in DMSO (1 mL) and as bulk solid quantities. Appearance is colorless to light yellow with a reported density of 1.164 g/cm3.
  • Endogenous metabolite useful as a reference standard for metabolic studies.
  • Available as ready-to-use 10 mM solution in DMSO for rapid use.
  • Also available in multiple powder sizes for bulk applications.
  • Documentation available including datasheet, SDS, COA, and spectral data.
  • Colorless to light yellow liquid with reported density 1.164 g/cm3.