Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-methoxy-5-oxopentanoic acid | 1501-27-5 | MFCD00004409 | 99.3% | 146.14 g/mol | C6H10O4 | 25 G
SDP

Catalog No. 5000303072
Encompass_Preferred

5-Methoxy-5-oxopentanoic acid is an endogenous metabolite supplied as a research-grade chemical standard for laboratory and analytical use. It is offered in solid and solution formats suitable for use as an analytical reference, metabolite standard, or reagent in biochemical and metabolomics studies.

  • Purity of 99.3%, suitable for analytical applications.
  • Molecular formula C6H10O4 and molecular weight 146.14 g/mol.
  • Available in multiple sizes and pre-dissolved DMSO solution formats.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
  • Colorless to light yellow liquid with density ~1.164 g/cm3.

Catalog No. 50-003-03072 Supplier Medchemexpress LLC Supplier No. 25GHYW017523
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Methoxy-5-oxopentanoic acid is an endogenous metabolite supplied as a research-grade chemical standard for laboratory and analytical use. It is offered in solid and solution formats suitable for use as an analytical reference, metabolite standard, or reagent in biochemical and metabolomics studies.

  • Purity of 99.3%, suitable for analytical applications.
  • Molecular formula C6H10O4 and molecular weight 146.14 g/mol.
  • Available in multiple sizes and pre-dissolved DMSO solution formats.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
  • Colorless to light yellow liquid with density ~1.164 g/cm3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.