Learn More
Medchemexpress LLC 5-methoxy-5-oxopentanoic acid | 1501-27-5 | MFCD00004409 | 99.3% | 146.14 g/mol | C6H10O4 | 5 G
SDP
Catalog No. 5000299983
Encompass_Preferred
5-Methoxy-5-oxopentanoic acid is an endogenous metabolite supplied as an analytical reference standard for biochemical and metabolic research. It is provided as a colorless to light-yellow liquid with formula C6H10O4 and molecular weight 146.14 g/mol, and is used for method development, quantitation, and quality control in metabolomics and analytical chemistry.
  • Endogenous metabolite used as an analytical standard.
  • Provided as a liquid, colorless to light yellow, with density ~1.164 g/cm3.
  • High purity suitable for analytical applications (reported ~99.3%).
  • Available in multiple pack sizes and solution formats for convenience.
  • Suitable for method development, calibration, and spike recovery studies.
  • Batch-specific analytical documentation (COA) provided for traceability.
Catalog No. 50-002-99983 Supplier Medchemexpress LLC Supplier No. HYW0175235G
Add to Cart
Add to Cart
5-Methoxy-5-oxopentanoic acid is an endogenous metabolite supplied as an analytical reference standard for biochemical and metabolic research. It is provided as a colorless to light-yellow liquid with formula C6H10O4 and molecular weight 146.14 g/mol, and is used for method development, quantitation, and quality control in metabolomics and analytical chemistry.
  • Endogenous metabolite used as an analytical standard.
  • Provided as a liquid, colorless to light yellow, with density ~1.164 g/cm3.
  • High purity suitable for analytical applications (reported ~99.3%).
  • Available in multiple pack sizes and solution formats for convenience.
  • Suitable for method development, calibration, and spike recovery studies.
  • Batch-specific analytical documentation (COA) provided for traceability.