Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Methoxy-a-ethyltry 10mM 1mL | 4765-10-0 | 218.29 g/mol | C13H18N2O | 1 ML
SDP

Catalog No. 5000308029
Encompass_Preferred

5-Methoxy-α-ethyltryptamine (5-MeO-α-ET; CAS 4765-10-0) is a tryptamine-class research compound supplied as a solid and as a 10 mM solution in DMSO for laboratory use. Molecular formula C13H18N2O; molecular weight 218.29 g/mol. For research use only; not for human consumption.

  • Chemical name: 5-methoxy-α-ethyltryptamine (CAS 4765-10-0).
  • Molecular formula: C13H18N2O.
  • Molecular weight: 218.29 g/mol.
  • Purity: 98.9% (as listed on COA).
  • Formulation: solid and 10 mM solution in DMSO (1 ml).
  • Storage: in solution store at -80°C up to 6 months or -20°C short term; protect from light.
  • Intended use: supplied for in vitro research applications.

Catalog No. 50-003-08029 Supplier Medchemexpress LLC Supplier No. HY11679510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

5-Methoxy-α-ethyltryptamine (5-MeO-α-ET; CAS 4765-10-0) is a tryptamine-class research compound supplied as a solid and as a 10 mM solution in DMSO for laboratory use. Molecular formula C13H18N2O; molecular weight 218.29 g/mol. For research use only; not for human consumption.

  • Chemical name: 5-methoxy-α-ethyltryptamine (CAS 4765-10-0).
  • Molecular formula: C13H18N2O.
  • Molecular weight: 218.29 g/mol.
  • Purity: 98.9% (as listed on COA).
  • Formulation: solid and 10 mM solution in DMSO (1 ml).
  • Storage: in solution store at -80°C up to 6 months or -20°C short term; protect from light.
  • Intended use: supplied for in vitro research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.