Learn More
Cayman Chemical 5-methoxy AMT 10mg
SDP
Catalog No. 502953483
Encompass_Preferred
A potent psychoactive analog of 5-methoxy DiPT and AMT that inhibits re-uptake (IC50s = 0.18, 2.9, and 3.37 μM) and stimulates release (EC50s = 1.5, 460, and 8.9 μM) of dopamine, serotonin, and norepinephrine, respectively from rat brain synaptosomes
Catalog No. 50-295-3483 Supplier Cayman Chemical Supplier No. 1155310 MG
A potent psychoactive analog of 5-methoxy DiPT and AMT that inhibits re-uptake (IC50s = 0.18, 2.9, and 3.37 μM) and stimulates release (EC50s = 1.5, 460, and 8.9 μM) of dopamine, serotonin, and norepinephrine, respectively from rat brain synaptosomes