Learn More
Apexbio Technology LLC 5-Methoxy-CTP(Synonyms: 5-Methoxycytidine triphosphate, 5-Methoxy-CTP, 5-MeO-CTP, 5-Methoxy Cytidine-5'-triphosphate), 10ul (100 mM).
SDP
Catalog No. 5000060736
Encompass_Preferred
5-Methoxy-CTP is a chemically modified cytidine triphosphate analog characterized by a methoxy group substitution at position 5 of the cytosine ring During in vitro transcription it can be enzymatically incorporated into synthesized RNA resulting in RNAs containing 5-methoxy-modified cytidine residues This methoxy substitution influences RNA chemical properties alters its structural characteristics and enhances resistance against intracellular nucleolytic degradation Consequently 5-Methoxy-CTP is valuable for investigating RNA structural dynamics RNA-protein interactions and RNA functional roles and it holds potential utility in RNA-based therapeutics functional genomics research and gene therapy approaches
Catalog No. 50-000-60736 Supplier Apexbio Technology LLC Supplier No. B80620.01
5-Methoxy-CTP is a chemically modified cytidine triphosphate analog characterized by a methoxy group substitution at position 5 of the cytosine ring During in vitro transcription it can be enzymatically incorporated into synthesized RNA resulting in RNAs containing 5-methoxy-modified cytidine residues This methoxy substitution influences RNA chemical properties alters its structural characteristics and enhances resistance against intracellular nucleolytic degradation Consequently 5-Methoxy-CTP is valuable for investigating RNA structural dynamics RNA-protein interactions and RNA functional roles and it holds potential utility in RNA-based therapeutics functional genomics research and gene therapy approaches