Learn More
Cayman Chemical 5-methoxy-DL-Tryptophan, Purity: ≥98%, CAS: 28052-84-8, 500 mg
SDP
Catalog No. 502991512
Encompass_Preferred
An active metabolite of tryptophan; reduces LPS-induced expression of COX-2, TNF-α, IL-1β, and IL-6 in RAW 264.7 macrophages at 50 mM; increases survival in a mouse model of LPS-induced endotoxemia and a mouse model of sepsis at 23.4 mg/kg; reduces tumor growth and the number of metastases in an A549 mouse xenograft model at 100 mg/kg
Catalog No. 50-299-1512 Supplier Cayman Chemical Supplier No. 31222500 MG
An active metabolite of tryptophan; reduces LPS-induced expression of COX-2, TNF-α, IL-1β, and IL-6 in RAW 264.7 macrophages at 50 mM; increases survival in a mouse model of LPS-induced endotoxemia and a mouse model of sepsis at 23.4 mg/kg; reduces tumor growth and the number of metastases in an A549 mouse xenograft model at 100 mg/kg