Learn More
Cayman Chemical 5-methoxy MiPT, Purity: ≥98%, CAS: 96096-55-8, 10 mg
SDP
Catalog No. 502955584
Encompass_Preferred
A psychedelic tryptamine which potently inhibits the re-uptake of the monoamines serotonin and norepinephrine (IC50s = 6.4 and 2.6 μM, respectively), but does not affect dopamine re-uptake; intended for forensic and research applications
Catalog No. 50-295-5584 Supplier Cayman Chemical Supplier No. 1148210 MG
A psychedelic tryptamine which potently inhibits the re-uptake of the monoamines serotonin and norepinephrine (IC50s = 6.4 and 2.6 μM, respectively), but does not affect dopamine re-uptake; intended for forensic and research applications