Learn More
Cayman Chemical 5-methoxy Tryptophol, Purity: ≥98%, CAS: 712-09-4, 50 mg
SDP
Catalog No. 502984166
Encompass_Preferred
A natural indole that is produced by the pineal gland
Catalog No. 50-298-4166 Supplier Cayman Chemical Supplier No. 2106150 MG
A natural indole that is produced by the pineal gland