Learn More
Apexbio Technology LLC 5-Methoxy-UTP 10x100ul (100 mM)
SDP
Catalog No. 503107241
Encompass_Preferred
5-Methoxy-UTP is a modified uridine triphosphate nucleotide bearing a methoxy group at the carbon-5 position This modification significantly alters the chemical properties and biological functions of the nucleotide During in vitro mRNA synthesis 5-Methoxy-UTP can be incorporated into RNA molecules generating RNAs containing 5-methoxyuridine residues Methoxy-modified RNA plays an important role in studies focusing on RNA structure function and interactions with proteins Additionally 5-Methoxy-UTP markedly improves RNA stability and protects against intracellular degradation Therefore this modification is widely utilized in gene expression research RNA drug development and gene therapy applications
Catalog No. 50-310-7241 Supplier Apexbio Technology LLC Supplier No. B80611
Add to Cart
Add to Cart
Request a Quote
5-Methoxy-UTP is a modified uridine triphosphate nucleotide bearing a methoxy group at the carbon-5 position This modification significantly alters the chemical properties and biological functions of the nucleotide During in vitro mRNA synthesis 5-Methoxy-UTP can be incorporated into RNA molecules generating RNAs containing 5-methoxyuridine residues Methoxy-modified RNA plays an important role in studies focusing on RNA structure function and interactions with proteins Additionally 5-Methoxy-UTP markedly improves RNA stability and protects against intracellular degradation Therefore this modification is widely utilized in gene expression research RNA drug development and gene therapy applications