Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-methoxyflavone | 42079-78-7 | MFCD00016942 | 100.0% | 252.26 g·mol⁻¹ | C16H12O3 | 1 ML
SDP

Catalog No. 5000242797
Encompass_Preferred

5-Methoxyflavone is a flavonoid-class small molecule (C16H12O3, MW 252.26 g·mol⁻¹) reported as a DNA polymerase-beta inhibitor and a neuroprotective agent. It is supplied as both a solid and a ready-to-use 10 mM solution in DMSO for in vitro research, with storage and solvent recommendations to support reproducible experiments.

  • High purity (99.97%) suitable for biochemical assays.
  • Available as a 10 mM solution in DMSO for ready-to-use applications.
  • Also supplied as solid in multiple pack sizes (25 mg-500 mg).
  • Molecular formula C16H12O3; molecular weight 252.26 g·mol⁻¹.
  • Recommended solvent: DMSO; may require sonication to dissolve.
  • Storage recommendations: powder -20°C for long term, solution -80°C for extended storage.

Catalog No. 50-002-42797 Supplier Medchemexpress LLC Supplier No. HY10779010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Methoxyflavone is a flavonoid-class small molecule (C16H12O3, MW 252.26 g·mol⁻¹) reported as a DNA polymerase-beta inhibitor and a neuroprotective agent. It is supplied as both a solid and a ready-to-use 10 mM solution in DMSO for in vitro research, with storage and solvent recommendations to support reproducible experiments.

  • High purity (99.97%) suitable for biochemical assays.
  • Available as a 10 mM solution in DMSO for ready-to-use applications.
  • Also supplied as solid in multiple pack sizes (25 mg-500 mg).
  • Molecular formula C16H12O3; molecular weight 252.26 g·mol⁻¹.
  • Recommended solvent: DMSO; may require sonication to dissolve.
  • Storage recommendations: powder -20°C for long term, solution -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.