Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-methoxyflavone | 42079-78-7 | MFCD00016942 | 100.0% | 252.26 g/mol | C16H12O3 | 100 MG
SDP

Catalog No. 5000242267
Encompass_Preferred

5-Methoxyflavone is a research-grade flavone (C16H12O3; CAS 42079-78-7) reported to inhibit DNA polymerase-β and exhibit neuroprotective activity. It is supplied as a high-purity solid intended for laboratory research and preparation of DMSO stock solutions.

  • High purity (reported 99.97%), suitable for analytical and biological studies.
  • Solid, white to off-white powder for convenient handling.
  • Soluble in DMSO for preparation of stock solutions.
  • Available in multiple pack sizes including 25 mg, 50 mg, 100 mg, and 500 mg.
  • Storage: powder stable at -20°C; in solvent store at -80°C for long-term stability.

Catalog No. 50-002-42267 Supplier Medchemexpress LLC Supplier No. HY107790100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Methoxyflavone is a research-grade flavone (C16H12O3; CAS 42079-78-7) reported to inhibit DNA polymerase-β and exhibit neuroprotective activity. It is supplied as a high-purity solid intended for laboratory research and preparation of DMSO stock solutions.

  • High purity (reported 99.97%), suitable for analytical and biological studies.
  • Solid, white to off-white powder for convenient handling.
  • Soluble in DMSO for preparation of stock solutions.
  • Available in multiple pack sizes including 25 mg, 50 mg, 100 mg, and 500 mg.
  • Storage: powder stable at -20°C; in solvent store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.