Learn More
Medchemexpress LLC 5-methoxyflavone | 42079-78-7 | MFCD00016942 | 100.0% | 252.26 g/mol | C16H12O3 | 50 MG
SDP
Catalog No. 5000242420
Encompass_Preferred
5-Methoxyflavone is a research-grade flavonoid compound used in biochemical and neuroscience studies. It acts as a DNA polymerase-beta inhibitor and is used in assays probing DNA/RNA synthesis and neuroprotective mechanisms.
  • High purity (99.97%) for reproducible experimental results.
  • Molecular weight 252.26 g/mol and formula C16H12O3 for accurate compound identification.
  • Available in small-scale quantities suitable for screening and assay development.
  • Applicable to studies of DNA/RNA synthesis inhibition and neuroprotection.
  • Supplied as a white to off-white powder, convenient for handling and formulation.
Catalog No. 50-002-42420 Supplier Medchemexpress LLC Supplier No. HY10779050MG
5-Methoxyflavone is a research-grade flavonoid compound used in biochemical and neuroscience studies. It acts as a DNA polymerase-beta inhibitor and is used in assays probing DNA/RNA synthesis and neuroprotective mechanisms.
  • High purity (99.97%) for reproducible experimental results.
  • Molecular weight 252.26 g/mol and formula C16H12O3 for accurate compound identification.
  • Available in small-scale quantities suitable for screening and assay development.
  • Applicable to studies of DNA/RNA synthesis inhibition and neuroprotection.
  • Supplied as a white to off-white powder, convenient for handling and formulation.