Learn More
Selleck Chemical LLC 5-methoxyflavone S4782-1g
SDP
Catalog No. 503109902
Encompass_Preferred
5-methoxyflavone is a novel DNA polymerase-beta inhibitor and neuroprotective agent against beta-amyloid toxicity
Catalog No. 50-310-9902 Supplier Selleck Chemical LLC Supplier No. S47821G
5-methoxyflavone is a novel DNA polymerase-beta inhibitor and neuroprotective agent against beta-amyloid toxicity