Learn More
Selleck Chemical LLC 5-methoxyflavone S4782-25mg
SDP
Catalog No. 503111971
Encompass_Preferred
5-methoxyflavone is a novel DNA polymerase-beta inhibitor and neuroprotective agent against beta-amyloid toxicity
Catalog No. 50-311-1971 Supplier Selleck Chemical LLC Supplier No. S478225MG
5-methoxyflavone is a novel DNA polymerase-beta inhibitor and neuroprotective agent against beta-amyloid toxicity