Learn More
Medchemexpress LLC HY-W007566 500mg Medchemexpress, 5-Methoxyindole-3-acetic acid CAS:3471-31-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW007566500MG
Encompass_Preferred
Medchemexpress, HY-W007566 500mg 5-Methoxyindole-3-acetic acid CAS:3471-31-6 5-Methoxyindole-3-acetic acid, isolated from pineal tissue, is a metabolite of Melatonin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3242