Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methoxypyrazine-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog No. p-7057297
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6682603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6682603 Supplier Thermo Scientific Chemicals Supplier No. H6682603

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40155-42-8
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD10697773
InChI Key YVGVOPNUEFTVQO-UHFFFAOYSA-N
Synonym 2-carboxy-5-methoxypyrazine, 2-carboxy-5-methoxy-1,4-diazine, 2-pyrazinecarboxylicacid, 5-methoxy, 5-methoxy-2-pyrazinecarboxylic acid, 2-pyrazinecarboxylic acid, 5-methoxy, 5-methoxy-pyrazine-2-carboxylic acid, 5-methyloxy-2-pyrazinecarboxylic acid, 5-methoxy-pyrazinecarboxylic acid, 5-methoxypyrazine-2-carboxylic acpubchem_ext_datasource_regid
PubChem CID 45789811
IUPAC Name 5-methoxypyrazine-2-carboxylic acid
SMILES COC1=NC=C(N=C1)C(=O)O

Specifications

Quantity 1 g
Beilstein 908499
Formula Weight 154.13
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Methoxypyrazine-2-carboxylic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.