Learn More
Indofine Chemical 5-Methoxyresorcin, 95%, 250 Mg
SDP
Supplier:  Indofine Chemical 011052 250 MG
Encompass_Preferred
5-METHOXYRESORCINOL 250 MG, Rare Organics & Biochemicals, 3,5-Dihydroxyanisole; Phloroglucinol monomethylether, C7H8O3, MW: 140.14, 0.95, 2174-64-3, MFCD00002285
Catalog No. 50-908-383